Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine monobromide, 98%, ACROS Organics™

Click to view available options
Quantity:
50g
250g
Packaging:
Glass bottle
7789-33-5
BrI
206.81
MFCD00011353
CBEQRNSPHCCXSH-UHFFFAOYSA-N
iodine monobromide, iodine bromide, iodobromine, iodine bromide ibr, unii-g0k622rd9n, iodinemonobromide, iodobromane, bromoiodine, jodmonobromid, hanus solution
82238
BrI
7789-33-5
BrI
206.81
MFCD00011353
CBEQRNSPHCCXSH-UHFFFAOYSA-N
iodine monobromide, iodine bromide, iodobromine, iodine bromide ibr, unii-g0k622rd9n, iodinemonobromide, iodobromane, bromoiodine, jodmonobromid, hanus solution
82238
BrI
C-X bond formation reagent
Assay Percent Range 97.5% min. (Iodometry)
Linear Formula IBr
Fieser 01,501; 02,224
Merck Index 15, 5059
Solubility Information Solubility in water: soluble. Other solubilities: solubel in alcohol,ether,carbon disulfide and,glacial acetic acid
Formula Weight 206.81
Physical Form Crystalline Mass, Crystals or Chunks
Percent Purity 98%
Density 4.4100g/mL
Packaging Glass bottle
Solubility Solubility in water: soluble. Other solubilities: solubel in alcohol, ether, carbon disulfide and, glacial acetic acid
Specific Gravity 4.41
Sulfated Ash 0.05% max.
Boiling Point 116.0°C
Color Black-Brown to Gray
Melting Point 42.0°C to 50.0°C
Quantity 250g
Chemical Name or Material Iodine monobromide
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 232-159-9