Learn More
Iodine monochloride, 1M solution in dichloromethane, Thermo Scientific Chemicals
$59.65 - $333.00
Specifications
Chemical Name or Material | Iodine monochloride |
---|---|
Name Note | 1, 0M solution in dichloromethane |
CAS | 7790-99-0, 75-09-2 |
Color | Brown to Orange |
Physical Form | Solution |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
AC370741000
|
Thermo Scientific Chemicals
370741000 |
100 mL | Glass bottle |
Each
for $59.65
Save 22% Reg :
Order in Bulk
Request bulk or custom formats |
|
||||
AC370748000
|
Thermo Scientific Chemicals
370748000 |
800 mL | Glass bottle |
Each for $333.00
Order in Bulk
Request bulk or custom formats |
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentSpecifications
Iodine monochloride | |
7790-99-0, 75-09-2 | |
Solution | |
ICl | |
01,502; 08,261; 10,212; 11,268 | |
iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french | |
QZRGKCOWNLSUDK-UHFFFAOYSA-N | |
iodochlorane | |
24640 | |
0.95 to 1.05M | |
1.42 |
1, 0M solution in dichloromethane | |
Brown to Orange | |
ClI | |
MFCD00011354 | |
15, 5060 | |
Solubility in water: insoluble | |
ClI | |
162.35 | |
162.35 | |
Glass bottle | |
1.4200g/mL |
Safety and Handling
GHS H Statement
Suspected of causing cancer.
Fatal if swallowed.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
GHS P Statement
Rinse mouth.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
R
GHS Signal Word: Danger
EINECSNumber : 232-236-7
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.