Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine monochloride, 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
7790-99-0
ClI
162.36
MFCD00011354
QZRGKCOWNLSUDK-UHFFFAOYSA-N
iodine monochloride,iodine chloride,iodochlorine,iodine chloride icl,chloroiodide,wijs' chloride,iodinemonochloride,chlorine iodide,unii-0smg5nlu45,protochlorure d'iode french
24640
ClI
7790-99-0
ClI
162.36
MFCD00011354
QZRGKCOWNLSUDK-UHFFFAOYSA-N
iodine monochloride,iodine chloride,iodochlorine,iodine chloride icl,chloroiodide,wijs' chloride,iodinemonochloride,chlorine iodide,unii-0smg5nlu45,protochlorure d'iode french
24640
ClI
C-X bond formation reagent
Assay Percent Range 97.5% min. (Iodometry)
Linear Formula ICl
Fieser 01,502; 08,261; 10,212; 11,268
Merck Index 15, 5060
Solubility Information Solubility in water: insoluble. Other solubilities: decomposes, soluble in ethanol, ether and cs2
Formula Weight 162.36
Physical Form Fuming Liquid
Percent Purity 98%
Density 3.2000g/mL
Packaging Glass bottle
Solubility Solubility in water: insoluble. Other solubilities: decomposes, soluble in ethanol, ether and cs2
Specific Gravity 3.2
Boiling Point 97.0°C
Color Red to Brown
Melting Point 26.0°C
Quantity 500g
Chemical Name or Material Iodine monochloride, 98%
Show More Show Less

GHS H Statement
Toxic in contact with skin.
Fatal if swallowed.
Causes severe skin burns and eye damage.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 232-236-7

RTECSNumber : NN1650000

TSCA : TSCA