Iodine monochloride, ACS, Alfa Aesar™

Moisture and Light Sensitive

Manufacturer: Alfa Aesar 4434836

Quantity 500g

 View more versions of this product

Catalog No. AA4434836

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7790-99-0
    Physical Form Crystalline/Liquid
    Molecular Formula ClI
    Formula Weight 162.36
    MDL Number MFCD00011354
    Synonym Chloroiodide
    Chemical Name or Material Iodine monochloride, For analysis ACS
    Grade ACS Grade

    Specifications

    Formula Weight 162.36
    Quantity 500g
    Color Red-black
    EINECS Number 232-236-7
    Physical Form Crystalline/Liquid
    CAS 7790-99-0
    Synonym Chloroiodide
    Molecular Formula ClI
    Appearance Red-black crystalline/liquid
    MDL Number MFCD00011354
    Health Hazard 1 H301-H311-H314-H318: Toxic if swallowed. Toxic in contact with skin. Causes severe skin burns and eye damage. Causes serious eye damage.
    Chemical Name or Material Iodine monochloride, For analysis ACS
    Grade ACS Grade

    H301-H311-H314-H318: Toxic if swallowed. Toxic in contact with skin. Causes severe skin burns and eye damage. Causes serious eye damage.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit