Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine monochloride solution, Macron Fine Chemicals™

Click to view available options
Quantity:
1L
4L
7790-99-0
ClI
162.35
MFCD00011354
QZRGKCOWNLSUDK-UHFFFAOYSA-N
iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french
24640
iodochlorane
ClI
7790-99-0
ClI
162.35
MFCD00011354
QZRGKCOWNLSUDK-UHFFFAOYSA-N
iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french
24640
iodochlorane
ClI
Formula Weight 162.35
Physical Form Liquid
Boiling Point 97.4°C
Color Red to Brown
Quantity 4L
Chemical Name or Material Iodine Monochloride Solution