Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Iodine Monochloride Solution, Wijs, for Iodine Absorption Number of Fats and Oils, Ricca Chemical
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
Packaging:
Amber Glass Bottle
Safety-coated Glass Bottle
Specifications
CAS Min % | 96.0, 1.41, 0.09 |
CAS Max % | 100.0, 1.46, 0.1 |
Packaging | Safety-coated Glass Bottle |
Quantity | 4 L |
Chemical Name or Material | Iodine Monochloride Solution, Wijs |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction