Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine Monochloride TS, (U.S.P. Test Solution), Spectrum™

Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Glass Bottle
7681-11-0,7647-01-0
IK
166.00
NLKNQRATVPKPDG-UHFFFAOYSA-M
potassium iodide
[K+].[I-]
7681-11-0,7647-01-0
IK
166.00
NLKNQRATVPKPDG-UHFFFAOYSA-M
potassium iodide
[K+].[I-]

I-166, 7790-99-0

Spectrum™ Iodine Monochloride TS, (U.S.P. Test Solution) is a solution, sometimes designated with "TS or Test Solution", that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.
CAS Max % 0.001,0.999
UN Number UN1789
Packaging Amber Glass Bottle
Quantity 500 mL