Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine, Resublimed p.a., ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass Bottle
7553-56-2
I2
253.809
PNDPGZBMCMUPRI-UHFFFAOYSA-N
iodine,diiodine,iodine crystals,iodine sublimed,tincture iodine,vistarin,eranol,iodio,iodine solution,iode
807
CHEBI:17606
molecular iodine
II
7553-56-2
I2
253.809
PNDPGZBMCMUPRI-UHFFFAOYSA-N
iodine,diiodine,iodine crystals,iodine sublimed,tincture iodine,vistarin,eranol,iodio,iodine solution,iode
807
CHEBI:17606
molecular iodine
II
C-X bond formation reagent
Additional Information Vapor Pressure: 0.41hPa at 25°C
Assay Percent Range 99.5%
Linear Formula I2
Fieser 01,495; 02,200; 03,159; 04,258; 05,346; 06,293; 07,179; 08,256; 09,248; 10,210; 11,261; 12,235; 13,133; 14,181; 15,172; 16,182; 17,153
Merck Index 15, 5057
Formula Weight 253.81
Physical Form Flakes or Spheres
Percent Purity ≥99.4% (Iodometry)
Grade Extra Pure
Chlorine and Bromine 0.005% max.
Density 8.8 (Air = 1.0)
Non_volatile Matter 0.01% max.
Packaging Glass Bottle
Sulfate 75ppm max.
Boiling Point 185.0°C
Color Gray
Melting Point 113°C
Quantity 2.5kg
Chemical Name or Material Iodine
Show More Show Less

GHS P Statement
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF INHALED: Remove to fresh and keep confortable for several minutes.

Warning