Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine, Sublimed (DEA List I Chemical), Crystal, Reagent, ACS, 99.8%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
6 x 25 g
2.5 kg
6 x 125 g
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
7553-56-2
I2
253.81
PNDPGZBMCMUPRI-UHFFFAOYSA-N
diiodine
II
7553-56-2
I2
253.81
PNDPGZBMCMUPRI-UHFFFAOYSA-N
diiodine
II

I1010, 7553-56-2, I2

Spectrum™ Iodine, Sublimed (DEA List I Chemical), Crystal, Reagent, ACS is a chemical element and an analytical reagent. As an ACS grade Reagent, Spectrum Chemical manufactured Iodine is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and purity. This item is a DEA List I Chemical and does require the completion of a Regulatory form.
CAS Max % 1
UN Number UN3495
Percent Purity 99.8%
Packaging Amber Glass Bottle
Quantity 6 x 25 g