Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine Tincture, 2% (w/v), USP, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
7681-82-5,64-17-5
INa
149.89
FVAUCKIRQBBSSJ-UHFFFAOYSA-M
sodium iodide
[Na+].[I-]
7681-82-5,64-17-5
INa
149.89
FVAUCKIRQBBSSJ-UHFFFAOYSA-M
sodium iodide
[Na+].[I-]

IO105, 7553-56-2

Spectrum™ Iodine Tincture, 2 Percent (w/v), USP is an antiseptic that is also referred to as weak iodine solution. Primarily used as an external disinfectant in emergency survival kits, as well as in operating rooms and hospitals, it can also be used to disinfect water of a suspicious nature for drinking purposes. Spectrum Chemical USP IODINE TINCTURE products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 0.02,0.98
UN Number UN1170
Packaging Amber Glass Bottle
Quantity 4 L