Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Iodine, Tincture Products for Science Education

Click to view available options
Quantity:
500mL
This item is not returnable. View return policy
This item is not returnable. View return policy
Concentration or Composition (by Analyte or Components) 40% Ethanol
Formula Weight 253.81
Packaging 500 mL PVC
Certificate of Analysis Certificates of Analysis are not available for Fisher Scientific™ education-grade chemicals. A hazardous surcharge will be applied to chemicals that are designated Hazardous by the U.S. Dept. of Transportation at the time of shipment.
Color Amber
Quantity 500mL
Chemical Name or Material Iodine, Tincture

ShelfLife : 36 months

DOTInformation : DOT Class 3, : Flammable Liquid