Learn More
Selleck Chemical LLCSupplier Diversity Partner Iodixanol DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S521825MG
Iodixanol is a nonionic hydrophilic compound commonly used as a contrast agent during coronary angiography.
Catalog No. 50-194-8350