Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Iodoacetic acid, 97+%

Manufacturer:  Alfa Aesar™ A1241014

Quantity 25g

 View more versions of this product

Catalog No. AAA1241014


  Check Availability
Add to cart

Specifications

Description

Applications
Iodoacetic acid is used as a reagent for the modification of sulfhydryl groups in organic synthesis. It reacts with cysteine moiety in proteins to prevent the re-formation of disulfide bonds during protein sequencing. It is also used as a general reagent in acylation reactions as well as in glycopeptide synthesis.

Solubility
Soluble in water, alcohol and ethanol. Slightly soluble in ether and chloroform.

Notes
Light sensitive. Store in cool place. Incompatible with strong oxidizing agents, bases and strong reducing agents.
Description & Specifications

Chemical Identifiers

64-69-7
185.95
UN2923
14,5025
ICH2CO2H
MFCD00002685
1739079
Iodoacetic acid

Specifications

64-69-7
185.95
UN2923
14,5025
79° to 82°C
Pungent
≥97%
ICH2CO2H
MFCD00002685
1739079
208°C (dec.)
25g
Iodoacetic acid

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H314
Toxic if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

RTECSNumber : AI3500000

TSCA : TSCA

Recommended Storage : 2°C to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit