Learn More
Iodoacetic acid, 97+%, Thermo Scientific Chemicals
Description
Iodoacetic acid is used as a reagent for the modification of sulfhydryl groups in organic synthesis. It reacts with cysteine moiety in proteins to prevent the re-formation of disulfide bonds during protein sequencing. It is also used as a general reagent in acylation reactions as well as in glycopeptide synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Iodoacetic acid is used as a reagent for the modification of sulfhydryl groups in organic synthesis. It reacts with cysteine moiety in proteins to prevent the re-formation of disulfide bonds during protein sequencing. It is also used as a general reagent in acylation reactions as well as in glycopeptide synthesis.
Solubility
Soluble in water, alcohol and ethanol. Slightly soluble in ether and chloroform.
Notes
Light sensitive. Store in cool place. Incompatible with strong oxidizing agents, bases and strong reducing agents.
Specifications
Melting Point | 79°C to 82°C |
Boiling Point | 208°C (decomposition) |
Odor | Pungent |
Quantity | 100 g |
UN Number | UN2923 |
Beilstein | 1739079 |
Sensitivity | Light sensitive |
Merck Index | 14,5025 |
Solubility Information | Soluble in water,alcohol and ethanol. Slightly soluble in ether and chloroform. |
Formula Weight | 185.95 |
Show More |
Safety and Handling
Hazard Category | H301-H314 |
Hazard Statement | GHS H Statement H301-H314 Toxic if swallowed. Causes severe skin burns and eye damage. |
Precautionary Statement | P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
DOTInformation | Hazard Class: 8; Packaging Group: I |
EINECSNumber | 200-590-1 |
RTECSNumber | AI3500000 |
TSCA | Yes |
Recommended Storage | Keep cold |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.