Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodoacetic Acid 97%, ACROS Organics™

Click to view available options
Quantity:
25g
50g
100g
Packaging:
Glass bottle
64-69-7
C2H3IO2
185.95
MFCD00002685
JDNTWHVOXJZDSN-UHFFFAOYSA-N
iodoacetic acid,acetic acid, iodo,iodoacetate,monoiodoacetic acid,monoiodoacetate,monoiodine acetate,iodoaceticacid,iodoethanoic acid,acetic acid, 2-iodo,kyselina jodoctova
5240
CHEBI:74571
2-iodoacetic acid
C(C(=O)O)I
64-69-7
C2H3IO2
185.95
MFCD00002685
JDNTWHVOXJZDSN-UHFFFAOYSA-N
iodoacetic acid,acetic acid, iodo,iodoacetate,monoiodoacetic acid,monoiodoacetate,monoiodine acetate,iodoaceticacid,iodoethanoic acid,acetic acid, 2-iodo,kyselina jodoctova
5240
CHEBI:74571
2-iodoacetic acid
C(C(=O)O)I
Quantity 100g
Color Brown to Yellow
Packaging Glass bottle
Melting Point 79.0°C to 83.0°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Linear Formula ICH2CO2H
Beilstein 02, 222
Merck Index 15, 5068
Solubility Information Solubility in water: 600g/L (20 c). Other solubilities: soluble in alcohol,slightly soluble in ether
Formula Weight 185.95
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Iodoacetic acid, 97%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Toxic if swallowed.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 200-590-1

RTECSNumber : AI3500000

TSCA : TSCA