Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodoacetonitrile, 95%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
624-75-9
C2H2IN
166.95
MFCD00001886
VODKOOOHHCAWFR-UHFFFAOYSA-N
iodoacetonitrile,acetonitrile, iodo,iodo-acetonitrile,2-iodoethanenitrile,2-iodo acetonitrile,2-iodoaceto-nitrile,2-iodanylethanenitrile,pubchem12543,acmc-1ba3n
69356
2-iodoacetonitrile
C(C#N)I
624-75-9
C2H2IN
166.95
MFCD00001886
VODKOOOHHCAWFR-UHFFFAOYSA-N
iodoacetonitrile,acetonitrile, iodo,iodo-acetonitrile,2-iodoethanenitrile,2-iodo acetonitrile,2-iodoaceto-nitrile,2-iodanylethanenitrile,pubchem12543,acmc-1ba3n
69356
2-iodoacetonitrile
C(C#N)I
Boiling Point 182°C to 184°C (720.0mmHg)
Flash Point 86°C
Specific Gravity 2.3
Refractive Index 1.5740 to 1.578
Quantity 25mL
Color Red to Brown
Infrared Spectrum Authentic
Density 2.3000g/mL
Packaging Glass bottle
Assay Percent Range 95%
Linear Formula ICH2CN
Beilstein 02, 223
Formula Weight 166.95
Physical Form Liquid
Percent Purity ≥94%
Chemical Name or Material Iodoacetonitrile
Show More Show Less

GHS H Statement:
Toxic if inhaled.
Toxic if swallowed.
Toxic in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger