Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodobenzene diacetate, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
3240-34-4
C10H11IO4
322.09
ZBIKORITPGTTGI-UHFFFAOYSA-N
iodobenzene diacetate,diacetoxyiodo benzene,iodosobenzene diacetate,phenyliodine diacetate,phenyliodo diacetate,phenyliodoso acetate,phenyliodosodiacetate,phenyliodoso diacetate,bis acetato phenyliodine,benzene, diacetoxyiodo
76724
[acetyloxy(phenyl)-$l^{3}-iodanyl] acetate
CC(=O)OI(C1=CC=CC=C1)OC(=O)C
3240-34-4
C10H11IO4
322.09
ZBIKORITPGTTGI-UHFFFAOYSA-N
iodobenzene diacetate,diacetoxyiodo benzene,iodosobenzene diacetate,phenyliodine diacetate,phenyliodo diacetate,phenyliodoso acetate,phenyliodosodiacetate,phenyliodoso diacetate,bis acetato phenyliodine,benzene, diacetoxyiodo
76724
[acetyloxy(phenyl)-$l^{3}-iodanyl] acetate
CC(=O)OI(C1=CC=CC=C1)OC(=O)C
Oxidation reagent
Quantity 100g
Infrared Spectrum Authentic
Melting Point 161.0°C to 165.0°C
Packaging Glass bottle
Color Colorless to Yellow
Loss on Drying 1% max. (60°C)
Assay Percent Range 97.5% min. (Iodometry)
Linear Formula C6H5I(O2CCH3)2
Fieser 01,508; 03,166; 04,266; 10,214; 13,242
Solubility Information Solubility in water: insoluble
Formula Weight 322.09
Physical Form Solution
Percent Purity 98%
Chemical Name or Material Iodobenzene diacetate
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 221-808-1