Alfa Aesar™ Iodocyclohexane, 98%, stab. with copper

Manufacturer: Alfa Aesar™ B2484036

Quantity 500g

 View more versions of this product

Catalog No. AAB2484036

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 626-62-0
    Molecular Formula C6H11I
    Formula Weight 210.06
    MDL Number MFCD00003826
    Beilstein 1900797
    Synonym Cyclohexyl iodide
    Chemical Name or Material Iodocyclohexane, stabilized with copper

    Specifications

    Formula Weight 210.06
    Boiling Point 180°C
    Flash Point 71°C (159°F)
    TSCA TSCA
    Molecular Formula C6H11I
    MDL Number MFCD00003826
    CAS 626-62-0
    Quantity 500g
    Beilstein 1900797
    Synonym Cyclohexyl iodide
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Chemical Name or Material Iodocyclohexane, stabilized with copper
    Assay 98%

    Description

    Applications
    Iodocyclohexane has been employed as reagent in demethylation of aryl methyl ethers in DMF under reflux condition.

    Solubility
    Soluble in ethanol, ether, acetone.

    Notes
    Light sensitive. Store away from oxidizing agent, light.

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit