Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Iodocyclohexane, 98%, stab. with copper

Click to view available options
Quantity:
25g
100g
500g
626-62-0
C6H11I
210.058
MFCD00003826
FUCOMWZKWIEKRK-UHFFFAOYSA-N
cyclohexane, iodo,cyclohexyl iodide,cyclohexyliodide,iodo-cyclohexane,1-iodocyclohexane,heptynyltrimethylsilane,iodocyclohexane,acmc-1att4,c-c6h11i
12289
iodocyclohexane
C1CCC(CC1)I
626-62-0
C6H11I
210.058
MFCD00003826
FUCOMWZKWIEKRK-UHFFFAOYSA-N
cyclohexane, iodo,cyclohexyl iodide,cyclohexyliodide,iodo-cyclohexane,1-iodocyclohexane,heptynyltrimethylsilane,iodocyclohexane,acmc-1att4,c-c6h11i
12289
iodocyclohexane
C1CCC(CC1)I
Applications
Iodocyclohexane has been employed as reagent in demethylation of aryl methyl ethers in DMF under reflux condition.

Solubility
Soluble in ethanol, ether, acetone.

Notes
Light sensitive. Store away from oxidizing agent, light.
Flash Point 71°C (159°F)
Boiling Point 180°C
Quantity 25g
Beilstein 1900797
Formula Weight 210.06
Percent Purity 98%
Chemical Name or Material Iodocyclohexane, stabilized with copper

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

TSCA : TSCA