Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodoethane, 98%, pure, stabilized with silver, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
75-03-6,64-19-7
C2H5I
155.97
MFCD00001091
HVTICUPFWKNHNG-UHFFFAOYSA-N
mercury 2+ ion acetic acid
15337
CHEBI:33211
iodoethane
CCI
75-03-6,64-19-7
C2H5I
155.97
MFCD00001091
HVTICUPFWKNHNG-UHFFFAOYSA-N
mercury 2+ ion acetic acid
15337
CHEBI:33211
iodoethane
CCI
Alkylation reagent
Viscosity 0.592 mPa.s (20°C)
Stabilizer silver
Packaging Glass bottle
Refractive Index 1.5100 to 1.5130
Specific Gravity 1.95
Infrared Spectrum Authentic
Flash Point 62°C
Quantity 500g
Melting Point -108.0°C
Color Colorless to Yellow or Pink
Boiling Point 69.0°C to 73.0°C
Density 1.9500g/mL
Assay Percent Range 97.5% min. (GC)
Beilstein 01,96
Merck Index 15,3868
Solubility Information Solubility in water: 4g/L (20°C),slowly decomposes in water. Other solubilities: miscible with most other organic solvents,soluble in alcohol,ether,chloroform
Formula Weight 155.97
Physical Form Liquid
Percent Purity 98%
Grade Pure
Chemical Name or Material Iodoethane, Stabilized
Show More Show Less

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing dif

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue

GHS Signal Word: Danger

EINECSNumber : 200-833-1

RTECSNumber : KI4750000

TSCA : TSCA

Recommended Storage : Product darkens on exposure to light