Alfa Aesar™ Iodomethane, 2M soln. in tert-butyl methyl ether

Manufacturer: Alfa Aesar™ H32660AE

Quantity 100mL

 View more versions of this product

Catalog No. AAH32660AE

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 74-88-4
    Molecular Formula CH3I
    Formula Weight 141.94
    MDL Number MFCD00001073
    UN Number UN1992
    Chemical Name or Material Iodomethane

    Specifications

    Formula Weight 141.94
    MDL Number MFCD00001073
    Concentration 2M soln. in tert-butyl methyl ether
    UN Number UN1992
    Boiling Point 41° to 43°C
    Quantity 100mL
    TSCA TSCA
    Molecular Formula CH3I
    CAS 74-88-4
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H301-H331-H351-H315-H335
    Highly flammable liquid and vapor.
    Toxic if swallowed.
    Toxic if inhaled.
    Suspected of causing cancer.
    Causes skin irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage 2° to 8°C
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
    Chemical Name or Material Iodomethane

    Description

    Applications
    Iodomethane is used to control insects, plant parasitic nematodes, soil borne pathogens and weed seeds. Iodomethane as a potential metal mobilizing agent in nature. Microhydration effects on the intermediates of the S(N)2 reaction of iodide anion with methyl iodide.

    Solubility
    Insoluble in water.

    Notes
    Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat, light, bases.

    GHS H Statement
    H225-H301-H331-H351-H315-H335
    Highly flammable liquid and vapor.
    Toxic if swallowed.
    Toxic if inhaled.
    Suspected of causing cancer.
    Causes skin irritation.
    May cause respiratory irritation.

    2° to 8°C

    GHS P Statement
    P210-P261-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit