Iodomethane, 99+%, stab. with copper, Alfa Aesar™

Manufacturer: Alfa Aesar 43681A1

Quantity 1kg

 View more versions of this product

Catalog No. AA43681A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 74-88-4
    Molecular Formula CH3I
    Formula Weight 141.94
    MDL Number MFCD00001073
    UN Number UN2644
    Beilstein 969135
    Merck Index 14,6087
    Synonym Methyl iodide
    Chemical Name or Material Iodomethane, stab. with copper

    Specifications

    Formula Weight 141.94
    Quantity 1kg
    Boiling Point 41° to 43°C
    CAS 74-88-4
    Merck Index 14,6087
    Synonym Methyl iodide
    Health Hazard 2 GHS H Statement
    H301-H331-H351-H312-H315-H335;
    Toxic if swallowed.
    Toxic if inhaled.
    Suspected of causing cancer.
    Harmful in contact with skin.
    Causes skin irritation.
    May cause respiratory irritation.
    Odor Pungent
    Health Hazard 3 GHS P Statement
    P261-P280-P281-P301+P310-P405-P501a;
    Avoid breathing dust/fume/gas/mist/vapours/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Sensitivity Light Sensitive
    TSCA TSCA
    Melting Point −67°C
    Beilstein 969135
    Color Colorless to Pale Yellow
    DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: METHYL IODIDE
    Physical Form Liquid
    Molecular Formula CH3I
    MDL Number MFCD00001073
    UN Number UN2644
    Health Hazard 1 Danger
    Storage Note 1 2° to 8°C
    RTECS Number PA9450000
    Chemical Name or Material Iodomethane, stab. with copper
    Assay ≥99%

    GHS H Statement
    H301-H331-H351-H312-H315-H335;
    Toxic if swallowed.
    Toxic if inhaled.
    Suspected of causing cancer.
    Harmful in contact with skin.
    Causes skin irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P281-P301+P310-P405-P501a;
    Avoid breathing dust/fume/gas/mist/vapours/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    2° to 8°C

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit