Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (Iodomethyl)cyclopropane, 97%

Manufacturer:  Alfa Aesar™ H35309MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH35309MD


  Check Availability
Add to cart

Specifications

Description

Applications
(Iodomethyl)cyclopropane is used as pharmaceutical intermediate.

Notes
Light sensitive. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

33574-02-6
182
(Iodomethyl)cyclopropane
C4H7I
MFCD06410734

Specifications

33574-02-6
182
250mg
97%
C4H7I
MFCD06410734
(Iodomethyl)cyclopropane

Safety and Handling

SDS & Safety Documents

GHS H Statement
H319-H317
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P305+P351+P338-P363-P321-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wash contaminated clothing before reuse.
Specific treatment (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit