Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodoquinol, Powder, USP, 96-100.5%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail
83-73-8
C9H5I2NO
396.95
UXZFQZANDVDGMM-UHFFFAOYSA-N
5,7-diiodoquinolin-8-ol
OC1=C2N=CC=CC2=C(I)C=C1I
83-73-8
C9H5I2NO
396.95
UXZFQZANDVDGMM-UHFFFAOYSA-N
5,7-diiodoquinolin-8-ol
OC1=C2N=CC=CC2=C(I)C=C1I

IO115, 83-73-8, C9H5I2NO

Spectrum™ Iodoquinol, Powder, USP Also known as Diquinol and iodoxin, is used in the treatment of amoebiasis and as a luminal amebicide. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical manufactured USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Fiber Drum
Ignition Residue 0.005
Loss on Drying 0.005
Quantity 25 kg
CAS Max % 1
Percent Purity 96 to 100.5%