Learn More
Iodotrimethylplatinum(IV), Thermo Scientific Chemicals
Description
They are used as catalysts in organic reactions. It is used in chemical, organic Intermediates research. They are used as platinum group compounds for the preparation of materials suitable for industrial applications, mainly in electronics as coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Orange to White |
Physical Form | Crystalline |
Quantity | 0.5 g |
Linear Formula | (CH3)3PtI |
Solubility Information | It is insoluble in water.It is sparingly soluble ether,alcohol,acetone or light petroleum. |
Formula Weight | 367.09 |
Odor | Odorless |
Sensitivity | Air & Light Sensitive |
Chemical Name or Material | Iodotrimethylplatinum(IV) |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
TSCA | No |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.