Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodotrimethylsilane, 95-97%, stabilized, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
100mL
250mL
Packaging:
Glass bottle
16029-98-4
C3H9ISi
200.1
MFCD00001028
CSRZQMIRAZTJOY-UHFFFAOYSA-N
trimethylsilyl iodide,trimethyliodosilane,silane, iodotrimethyl,iodo trimethyl silane,trimethylsilyliodide,tms-iodide,iodotrimethysilane,iodotrimetylsilane,tms iodide,iodo trimethylsilane
85247
iodo(trimethyl)silane
C[Si](C)(C)I
16029-98-4
C3H9ISi
200.1
MFCD00001028
CSRZQMIRAZTJOY-UHFFFAOYSA-N
trimethylsilyl iodide,trimethyliodosilane,silane, iodotrimethyl,iodo trimethyl silane,trimethylsilyliodide,tms-iodide,iodotrimethysilane,iodotrimetylsilane,tms iodide,iodo trimethylsilane
85247
iodo(trimethyl)silane
C[Si](C)(C)I
C-X bond formation reagent
Stabilizer copper
Specific Gravity 1.4
Flash Point −31°C
Packaging Glass bottle
Color Colorless to Red
Density 1.4000g/mL
Quantity 5mL
Refractive Index 1.4700 to 1.4800
Boiling Point 106.0°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (Ex I) (Argentometry)
Linear Formula (CH3)3SiI
Fieser 08,261; 09,251; 10,216; 11,271; 12,259; 13,329; 16,188; 06,81
Solubility Information Solubility in water: reacts
Formula Weight 200.1
Physical Form Liquid
Percent Purity 95-97%
Chemical Name or Material Iodotrimethylsilane, 95-97%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 240-171-0

TSCA : TSCA