Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ion exchanger Dowex™ 50 WX 4, H+ form, EMD-Millipore

Click to view available options
Quantity:
250g
69011-20-7
C28H30
366.548
NWUYHJFMYQTDRP-UHFFFAOYSA-N
porapak p, lewatit c267 ion exchange resin, ion exchange resin, benzene, diethenyl-, polymer with ethenylbenzene and ethenylethylbenzene, cation exchange resin, ethidium bromide adsorber, divinylbenzene, ethylvinylbenzene, vinylbenzene copolymer, ethenylbenzene, diethenylbenzene, ethylethenylbenzene polymer, ethylbenzene, ethylethenylbenzene, diethenylbenzene polymer, sulfonated, benzene, diethenyl-, polymer with ethenylbenzene and ethenylethylbenzene, sulfonated
172244
1,2-bis(ethenyl)benzene;1-ethenyl-2-ethylbenzene;styrene
CCC1=CC=CC=C1C=C.C=CC1=CC=CC=C1.C=CC1=CC=CC=C1C=C
69011-20-7
C28H30
366.548
NWUYHJFMYQTDRP-UHFFFAOYSA-N
porapak p, lewatit c267 ion exchange resin, ion exchange resin, benzene, diethenyl-, polymer with ethenylbenzene and ethenylethylbenzene, cation exchange resin, ethidium bromide adsorber, divinylbenzene, ethylvinylbenzene, vinylbenzene copolymer, ethenylbenzene, diethenylbenzene, ethylethenylbenzene polymer, ethylbenzene, ethylethenylbenzene, diethenylbenzene polymer, sulfonated, benzene, diethenyl-, polymer with ethenylbenzene and ethenylethylbenzene, sulfonated
172244
1,2-bis(ethenyl)benzene;1-ethenyl-2-ethylbenzene;styrene
CCC1=CC=CC=C1C=C.C=CC1=CC=CC=C1.C=CC1=CC=CC=C1C=C

Strongly acid cation exchangers

Ignition Point >500°C
Water Absorption 640 to 720g/kg
Quantity 250g
Chemical Name or Material Ion exchanger Dowex™ 50 WX 4

Recommended Storage : Store at 5° to 30°C