Alfa Aesar™ Ionac™ NM-65, Ion Exchange Resin, Indicator dyed

Production of water of low, total dissolved solids, low CO2, and low silica

Manufacturer: Alfa Aesar™ 4370536

Quantity 500g

 View more versions of this product

Catalog No. AA4370536

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    MDL Number MFCD00145887
    Chemical Name or Material Ionac™ NM-65 Ion Exchange Resin, Indicator dyed

    Specifications

    TSCA TSCA
    Physical Form Beads
    MDL Number MFCD00145887
    Quantity 500g
    Chemical Name or Material Ionac™ NM-65 Ion Exchange Resin, Indicator dyed

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit