Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ionic Strength Adjustor, pH Indicating for use with Ammonia Selective Electrode, Reagents

Catalog No. p-200021718
Click to view available options
Quantity:
500 mL
1 L

Ionic Strength Adjustor, pH Indicating for use with Ammonia Selective Electrode, Reagent, use for Ammonia analysis. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5, 1310-73-2, 67-56-1
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Packaging Poly Bottle
Quantity 1 L
Specific Gravity 1.1205
Physical Form Liquid
Color Dark Blue
pH 14
Boiling Point 103°C
For Use With (Equipment) Water Analyzer
Solubility Completely miscible
Grade Reagent
Chemical Name or Material Ionic Strength Adjustor, pH Indicating for use with Ammonia Selective Electrode
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.