Learn More
Ionic Strength Adjustor, pH Indicating and Adjusting, for use with Ammonia Selective Electrode, Ricca Chemical
Supplier: Ricca Chemical Company 412832
Specifications
Ionic Strength Adjustor | |
65.0, 26.25, 5.14, 1.22, 0.01 | |
1310-73-2, 67-56-1, 6381-92-6, 125-20-2 | |
1 L |
pH Indicating and Adjusting, for use with Ammonia Selective Electrode | |
68.0, 27.32, 5.35, 1.27, 0.01 | |
Natural Poly Bottle |