Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SI Analytics™ Ionselective Indicator Electrode
For YSI TitroLine Autotitrators
Supplier: SI Analytics™ 285216312
Description
- Copper Junction
- pH Range: 2 to 6
Specifications
Ag/AgCl | |
Plastic | |
0°C to 80°C |
1 m | |
Cu 1100 A | |
Ion Selective Indicator Electrode |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction