Learn More
Biocare Medical, LLCSupplier Diversity Partner IP BACKGROUND PUNISHER 20ML  DFS Item
Manufacturer:  Biocare Medical, LLCSupplier Diversity Partner IP974G20
intelliPATH Background Punisher is a universal blocking reagent used for reducing nonspecific IHC background staining. It is specially formulated for use on an intelliPATH Automated Stainer with superior pH stability.
Catalog No. 50-719-797