Learn More
Brady™ IP™ Thermal Transfer Printer: PROMO
Fully integrated, easy-to-use printer, material, and software system.
Manufacturer:  Brady™ BPIP600CPPM30
 View more versions of this product
Catalog No. 19-104-499PM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Smart Cell technology allows the printer, materials, and ribbon to communicate with each other. The system offers material recognition, automatic formatting, and user-friendly printer features for labels, sleeves, and tags.
  • Brady IP™ Printer detects loaded material and ribbon and displays the user information on the LCD
  • The material information is sent to the software, where template and settings are automatically populated
  • Create data on template, then click and print.
Specifications
Specifications
Printer
Serial, USB 2.0 and Ethernet 10/100
Barcode labeling, circuit board labeling, control panel/push button, data communications labeling, DoD UID, laboratory identification, product identification, wire and cable marking, wire and cable marking - ADM
64MB
AC power and USB, serial and ethernet 10/100 connectivity
Thermal Transfer
13 lb.
5 Years
Single Color Printing
Graphical and Alphanumeric LCD
Cutter
8 hrs / 5 days
up to 4 in./Sec
600dpi
4.160 in.