Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IP6K1 Mouse anti-Human, Biotin, Clone: 10611, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243667B

Catalog No. NBP243667B


Add to cart

Description

Description

IP6K1 Monoclonal antibody specifically detects IP6K1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

IP6K1
10611
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
IP6K1
Recombinant protein encompassing a sequence within the center region of human IP6K1. The exact sequence is proprietary.
Protein G purified
Neuroscience, Neurotransmission, Signal Transduction
Primary
9807
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Biotin
PBS with 0.05% Sodium Azide
ATP:1D-myo-inositol-hexakisphosphate phosphotransferase, EC 2.7.4.21, inositol hexakisphosphate kinase 1, Inositol hexaphosphate kinase 1KIAA0263IHPK1PiUS, InsP6 kinase 1, MGC9925, Pi uptake stimulator
Mouse
IgG2b
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit