Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IP6K1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP259105
Description
IP6K1 Polyclonal specifically detects IP6K1 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
IP6K1 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
ATP:1D-myo-inositol-hexakisphosphate phosphotransferase, EC 2.7.4.21, inositol hexakisphosphate kinase 1, Inositol hexaphosphate kinase 1KIAA0263IHPK1PiUS, InsP6 kinase 1, MGC9925, Pi uptake stimulator | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide | |
IP6K1 | |
Maps to a region between 75 and 125 | |
100 μL | |
Neuroscience, Neurotransmission, Signal Transduction | |
9807 | |
Store at 4C. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction