Learn More
IP6K2 Mouse anti-Human, Clone: OTI1F12, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF808811
Catalog No. 50-166-9093
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Inositol 1,4,5-trisphosphate (IP3) is a second messenger for many growth factors, hormones, and neurotransmitters and, upon binding to the IP3 receptor (IP3R), triggers the release of intracellular, luminal calcium to the cytosol. Several additional cellular forms of inositol phosphates have been discovered. These include some which have been recently discovered and characterized to possess up to seven or eight phosphate groups, including pyrophosphates (PP-IPs: pyrophosphate-inositol phosphates). It has been proposed that high-energy pyrophosphates may participate in protein phosphorylation. Inositol-hexakisphosphate (IP6) kinase, or PiUS (Pi Uptake Stimulator) has ATP synthase activity and transfers a phosphate from PP-IP5 to ADP to form ATP thus suggesting an important physiological role for PP-IP5 and IP6 kinase.
Specifications
Specifications
IP6K2
Monoclonal
Unconjugated
IP6K2
Lyophilized
IP6K2
Human recombit protein fragment corresponding to amino acids 1-250 of human IP6K2 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI1F12
PBS with 8% trehalose and no preservative; pH 7.3
Q9UHH9
IHPK2, PIUS
Mouse
IgG2a
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
51447