Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IP6K2 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549476

Catalog No. PA549476


Add to cart

Description

Description

Inositol 1,4,5-trisphosphate (IP3) is a second messenger for many growth factors, hormones, and neurotransmitters and, upon binding to the IP3 receptor (IP3R), triggers the release of intracellular, luminal calcium to the cytosol. Several additional cellular forms of inositol phosphates have been discovered. These include some which have been recently discovered and characterized to possess up to seven or eight phosphate groups, including pyrophosphates (PP-IPs: pyrophosphate-inositol phosphates). It has been proposed that high-energy pyrophosphates may participate in protein phosphorylation. Inositol-hexakisphosphate (IP6) kinase, or PiUS (Pi Uptake Stimulator) has ATP synthase activity and transfers a phosphate from PP-IP5 to ADP to form ATP thus suggesting an important physiological role for PP-IP5 and IP6 kinase.
Specifications

Specifications

IP6K2
Polyclonal
PBS with 0.09% sodium azide
Q80V72
Inositol hexakisphosphate kinase 2, InsP6 kinase 2, P(i)-uptake stimulator, PiUS, IP6K2, IHPK2
Rabbit
IgG
200 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
76500
Western Blot
Unconjugated
Ip6k2
Liquid
IP6K2
KLH conjugated synthetic peptide between 226-258 amino acids from the Central region of human IP6K2.
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit