Learn More
Tocris Bioscience™ IP7e Available on GSA/VA Contract for Federal Government customers only.
Potent Nurr1 activator
$178.80 - $748.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
569910 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5699/10
Each for $178.80
Add to cart
569950 View Documents Tocris Bioscience™
5699/50
Each for $748.00
Add to cart
Description
Description
Potent Nurr1 activator (EC50 = 3.9 nM). Delays onset and reduces severity of symptoms in mice with experimental autoimmune encephalomyelitis (EAE). Also suppresses NF-κB signaling. Brain penetrant and orally bioavailable.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.