Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Anti- Ipilimumab (Anti-Idiotype) Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems MAB10064100
This item is not returnable.
View return policy
Description
Ipilimumab Monoclonal specifically detects Ipilimumab in Multi-Species samples. It is validated for Western Blot.Specifications
Ipilimumab | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Full length Ipilimumab | |
100 μg | |
Detects Ipilimumab in direct ELISAs and Western blots. In Western blots, no cross-reactivity with Cetuximab, Brentuximab, Teprotumumab, and Aducanumab is observed. | |
Multi-species | |
Supernatant |
Western Blot | |
1001345 | |
Western Blot 2 μg/mL | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction