Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ipilimumab Mouse anti-Multi-Species, Clone: 1001345, R&D Systems™
Mouse Monoclonal Antibody
$136.67 - $456.67
Specifications
Antigen | Ipilimumab |
---|---|
Clone | 1001345 |
Dilution | Western Blot 2 μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10064SP
|
R&D Systems
MAB10064SP |
25 μg |
Each for $136.67
|
|
MAB10064100
|
R&D Systems
MAB10064100 |
100 μg |
Each for $456.67
|
|
Description
Ipilimumab Monoclonal specifically detects Ipilimumab in Multi-Species samples. It is validated for Western Blot.Specifications
Ipilimumab | |
Western Blot 2 μg/mL | |
Monoclonal | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
1001345 | |
Western Blot | |
Unconjugated | |
Many | |
Full length Ipilimumab | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects Ipilimumab in direct ELISAs and Western blots. In Western blots, no cross-reactivity with Cetuximab, Brentuximab, Teprotumumab, and Aducanumab is observed. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title