Learn More
BioworldSupplier Diversity Partner IPL-41 Insect Medium, 1 L DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 306110442
Developed in a series of media by a group from the United States Department of Agriculture (USDA) Insect Pathology Laboratory, IPL-41 is a modification of Goodwin's orginal IPL formulation. It was orginally designed to support the growth and maintenance of cell lines descendant from lepidopteran species and the propagation of viruses within these cells. It has been used for large scale production of baculovirus. IPL-41 Insect Medium (Powder), item #785095, contains L-glutamine and calcium chloride. Does not include tryptose phosphate broth and sodium bicarbonate.
Catalog No. 50-254-107