Learn More
IPO-38 Mouse anti-Human, Clone: IPO-38, Abnova™
Mouse monoclonal antibody raised against native human IPO-38.
Manufacturer:  Abnova Corporation MAB13232
Catalog No. 89-934-382
Add to cart
Description
Description
Specifications
Specifications
IPO-38
IPO-38
Mouse monoclonal antibody raised against native human IPO-38.
In 10mM PBS.
Mouse
IgM, kappa
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
Spleen cells of a patient with hairy cell leukemia.
Polyethylene Glycol (PEG) precipitation
RUO
Primary
Human