Learn More
IPO-38 Mouse anti-Human, Clone: SPM260, Abnova™
Mouse monoclonal antibody raised against native human IPO-38.
Manufacturer:  Abnova Corporation MAB13205
Catalog No. 89-934-383
Add to cart
Description
Description
Specifications
Specifications
IPO-38
SPM260
Mouse monoclonal antibody raised against native human IPO-38.
In 10mM PBS (0.05% BSA, 0.05% sodium azide).
Mouse
IgM, kappa
100 ug
Store at 4°C.
Monoclonal
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
Spleen cells of a patient with hairy cell leukemia.
PEG precipitation
RUO
Primary
Human