Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Importin 13 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550432

Catalog No. PA550432


Add to cart

Description

Description

The antibody detects endogenous levels of total IPO13 protein.

This gene encodes a member of the importin-beta family of nuclear transport proteins. The encoded protein mediates the import of specific cargo proteins from the cytoplasm to the nucleus and is dependent on the Ras-related nuclear protein-GTPase system. The encoded protein is also involved in nuclear export of the eukaryotic translation initiation factor 1A.
Specifications

Specifications

Importin 13
Polyclonal
Unconjugated
IPO13
Liquid
IPO13
Fusion protein corresponding to residues near the C terminal of human importin 13
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1.4 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
O94829
KIAA0724, RANBP13
Rabbit
IgG
100 μL
-20°C
Primary
9670
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit