Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SP110 Mouse anti-Human, Clone: OTI2A1, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526930

Catalog No. PIMA526930


Add to cart

Description

Description

Susceptibility to tuberculosis (TB) in mice has recently been attributed to the IPR1 gene. IPR1 is a member of the SP100/SP140 family of nuclear body proteins and encodes a leukocyte-specific nuclear body component. The protein can function as an activator of gene transcription and may serve as a nuclear hormone receptor coactivator. Alternative splicing has been observed for this gene and three transcript variants, encoding distinct isoforms, have been identified. SP110 is the closest homolog of the IPR1 protein in humans. The IPR1/Sp110 gene product might play a role in integrating signals generated by intracellular pathogens with mechanisms controlling innate immunity, cell death, and pathogenesis. IPR1/Sp110 is up-regulated after infection with M. tuberculosis and required by Anaplasma phagocytophilum for infection of human promyelocytic cells. Defects in Sp110 are a cause of severely impaired resistance to infection by M. tuberculosis.
Specifications

Specifications

SP110
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9HB58
IFI41, IFI75, IPR1, VODI; interferon-induced protein 41, 30kD; interferon-induced protein 41/75; interferon-induced protein 75, 52kD; phosphoprotein 41; phosphoprotein 75; sp110 nuclear body protein; speckled 110 kDa; transcriptional coactivator Sp110
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3431
Immunohistochemistry (Paraffin), Western Blot
OTI2A1
Unconjugated
SP110
Liquid
SP110
Human recombit protein fragment corresponding to amino acids 232-521 of SP110 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit