Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ipratropium Bromide, BP, EP, 99-100.5%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
Poly Pail
66985-17-9
C20H32BrNO4
430.38
KEWHKYJURDBRMN-XFQAGIBXSA-M
(1R,5S)-3-[(3-hydroxy-2-phenylpropanoyl)oxy]-8-methyl-8-(propan-2-yl)-8-azabicyclo[3.2.1]octan-8-ium hydrate bromide
O.[Br-].CC(C)[N+]1(C)[C@H]2CC[C@@H]1CC(C2)OC(=O)C(CO)C1=CC=CC=C1
66985-17-9
C20H32BrNO4
430.38
KEWHKYJURDBRMN-XFQAGIBXSA-M
(1R,5S)-3-[(3-hydroxy-2-phenylpropanoyl)oxy]-8-methyl-8-(propan-2-yl)-8-azabicyclo[3.2.1]octan-8-ium hydrate bromide
O.[Br-].CC(C)[N+]1(C)[C@H]2CC[C@@H]1CC(C2)OC(=O)C(CO)C1=CC=CC=C1

I1176, 66985-17-9, C20H30BrNO3•H2O

Spectrum™ Ipratropium Bromide, BP, EP is a bronchodilator that dilates airways in the lungs. All Spectrum Chemical manufactured BP, EP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Sulfated Ash 0.001
Optical Rotation -0.10° to +0.10°
Quantity 1 g
Water 3.9-4.4%
CAS Max % 1
Percent Purity 99 to 100.5%