Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IQ 1, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
331001-62-8
C21H22N4O2
362.433
ALJIEVIJBAJISI-NCELDCMTSA-N
iq 1,iq-1,2-4-acetyl-phenylazo-2-3,3-dimethyl-3,4-dihydro-2h-isoquinolin-1-ylidene-acetamide,2e-2-4-acetylphenyl hydrazinylidene-2-3,3-dimethyl-4h-isoquinolin-1-yl acetamide,2-4-acetylphenyl azo-2-3,4-dihydro-3,3-dimethyl-1 2h-isoquinolinylidene acetamide,2-4-acetylphenyl diazenyl-2-3,3-dimethyl 2,3,4-trihydroisoquinolylidene a cetamide,2-2-4-acetylphenyl diazenyl-2-3,4-dihydro-3,3-dimethyl-1 2h-isoquinolinylidene-acetamide
5823908
(2E)-2-[(4-acetylphenyl)hydrazinylidene]-2-(3,3-dimethyl-4H-isoquinolin-1-yl)acetamide
CC(=O)C1=CC=C(C=C1)NN=C(C2=NC(CC3=CC=CC=C32)(C)C)C(=O)N
This item is not returnable. View return policy
331001-62-8
C21H22N4O2
362.433
ALJIEVIJBAJISI-NCELDCMTSA-N
iq 1,iq-1,2-4-acetyl-phenylazo-2-3,3-dimethyl-3,4-dihydro-2h-isoquinolin-1-ylidene-acetamide,2e-2-4-acetylphenyl hydrazinylidene-2-3,3-dimethyl-4h-isoquinolin-1-yl acetamide,2-4-acetylphenyl azo-2-3,4-dihydro-3,3-dimethyl-1 2h-isoquinolinylidene acetamide,2-4-acetylphenyl diazenyl-2-3,3-dimethyl 2,3,4-trihydroisoquinolylidene a cetamide,2-2-4-acetylphenyl diazenyl-2-3,4-dihydro-3,3-dimethyl-1 2h-isoquinolinylidene-acetamide
5823908
(2E)-2-[(4-acetylphenyl)hydrazinylidene]-2-(3,3-dimethyl-4H-isoquinolin-1-yl)acetamide
CC(=O)C1=CC=C(C=C1)NN=C(C2=NC(CC3=CC=CC=C32)(C)C)C(=O)N

Enables Wnt/β-catenin-driven expansion of ESCs; prevents spontaneous differentiation

Maintains embryonic stem cell (ESC) pluripotency in Wnt3a-treated cells; enables Wnt/β-catenin-driven expansion of mouse ESCs and prevents spontaneous differentiation. Upregulates expression of Oct4 and Sox2 transcription factors. Binds to serine/threonine phosphatase PP2A and prevents PP2A/Nkd interaction.
Quantity 50mg
Formula Weight 362.42
Percent Purity >99%
Chemical Name or Material IQ 1

Recommended Storage : Store at -20°C