Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IQ 3, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective JNK3 inhibitor (Kd values are 66, 240 and 290 nM for JNK3, JNK1 and JNK2 respectively). Inhibits NF-κB/AP1 transcriptional activity in THP1-Blue cells (IC50 = 1.4 μM). Also inhibits TNF-α and IL-6 production in vitro.
Specifications
Color | Yellow |
Quantity | 50 mg |
Formula Weight | 341.32 |
Percent Purity | >98% |
Physical Form | Solid |
Chemical Name or Material | IQ 3 |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction