Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IQCA1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548321

Catalog No. PA548321

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The protein encoded by this gene is a member of the ATPases Associated with diverse cellular Activities (AAA) superfamily. Members of this superfamily, found in all organisms, participate in a large number of cellular processes and contain the ATPase module consisting of an alpha-beta-alpha core domain and the Walker A and B motifs of the P-loop NTPases. Alternative splicing results in multiple transcript variants that encode different protein isoforms. [provided by RefSeq, Jul 2012]
Specifications

Specifications

IQCA1
Polyclonal
PBS with 0.09% sodium azide
Q86XH1
IQ and AAA domain-containing protein 1, IQCA1, IQCA
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79781
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
IQCA1
Liquid
IQCA1
KLH conjugated synthetic peptide between 794-822 amino acids from the C-terminal region of human IQCA1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit