Learn More
Aviva Systems Biology Corp IQCE Antibody - middle region (ARP55486_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP55486_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IQCE. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human IQCE. Alternate names: 1700028P05Rik, KIAA1023, MGC41907ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-61522